Gene Transcripts

Gene Symbol
SH2D3A
Entrez Gene ID
10045
Gene Name
SH2 domain containing 3A
Gene Aliases
NSP1
Chromosome Location
Chr. 19: 6752160 - 6767513 on Build GRCh38
UniGene ID
Hs.439645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005490.2 NP_005481.2 4 627
XM_005259467.1 XP_005259524.1 4 577
XM_005259472.2 XP_005259529.1 3 190
XM_006722602.1 XP_006722665.1 4 577
XM_011527608.2 XP_011525910.1 3 994
XM_011527609.2 XP_011525911.1 3 491
XM_017026123.1 XP_016881612.1 4 577
XM_017026124.1 XP_016881613.1 4 577
GenBank mRNA AF124249.1 - 4 584
AK024368.1 - 3 1266
AK225386.1 - 4 550
AK290293.1 - 4 703
AK292781.1 - 4 550
AK299406.1 - 3 200
AK310659.1 - 4 550
AY358406.1 - 4 584
BC006281.2 - 4 564 3958342

View Full Product Details