Gene Transcripts
- Gene Symbol
- TEDDM1
- Entrez Gene ID
- 127670
- Gene Name
- transmembrane epididymal protein 1
- Gene Aliases
- EDDM9, Epdd1, HE9, HEL-S-45e, TMEM45C
- Chromosome Location
- Chr. 1: 182398117 - 182400616 on Build GRCh38
- UniGene ID
- Hs.156977
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172000.3 |
NP_741997.3
|
1 |
291 |
|
View Full Product Details