Gene Transcripts

Gene Symbol
MORC2
Entrez Gene ID
22880
Gene Name
MORC family CW-type zinc finger 2
Gene Aliases
CMT2Z, ZCW3, ZCWCC1
Chromosome Location
Chr. 22: 30925130 - 30968497 on Build GRCh38
UniGene ID
Hs.555918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303256.2 NP_001290185.1 5 703
NM_001303257.2 NP_001290186.1 5 703
NM_014941.3 NP_055756.1 6 1569
XM_011530004.1 XP_011528306.1 5 928
XM_017028667.1 XP_016884156.1 5 928
XM_017028668.1 XP_016884157.1 6 1023
GenBank mRNA AB020659.1 - 6 1483
BC019257.1 - 4 230 3503651
BC136782.1 - 6 428 9020772
CR456469.1 - 4 229
DR000665.1 - 5 651

View Full Product Details