Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100875.2 | NP_001094345.1 | 6 | 2138 | |
NM_001288779.1 | NP_001275708.1 | 7 | 2333 | ||
NM_001288780.1 | NP_001275709.1 | 7 | 2364 | ||
NM_152598.3 | NP_689811.2 | 6 | 2219 | ||
XM_005257095.2 | XP_005257152.2 | 7 | 2295 | ||
XM_005257096.2 | XP_005257153.2 | 7 | 2240 | ||
XM_005257097.3 | XP_005257154.2 | 8 | 2387 | ||
XM_005257098.2 | XP_005257155.2 | 7 | 2292 | ||
XM_005257100.2 | XP_005257157.2 | 6 | 2181 | ||
XM_005257102.2 | XP_005257159.2 | 6 | 2178 | ||
XM_005257103.1 | XP_005257160.1 | 6 | 2178 | ||
XM_005257106.4 | XP_005257163.2 | 6 | 1946 | ||
XM_011524435.1 | XP_011522737.1 | 7 | 2292 | ||
XM_011524436.1 | XP_011522738.1 | 6 | 2126 | ||
XM_011524437.1 | XP_011522739.1 | 7 | 2270 | ||
GenBank mRNA | AK093076.1 | - | 6 | 2172 | |
AK097506.1 | - | 1 | 1351 | ||
AK122947.1 | - | 7 | 2273 | ||
BC035021.1 | - | 6 | 2142 | 4824637 | |
BX648705.1 | - | 7 | 2296 |