Gene Transcripts

Gene Symbol
CLN8
Entrez Gene ID
2055
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
C8orf61, EPMR
Chromosome Location
Chr. 8: 1755778 - 1786570 on Build GRCh38
UniGene ID
Hs.127675
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018941.3 NP_061764.2 2 614
XM_005266021.3 XP_005266078.1 2 558
XM_005266022.1 XP_005266079.1 2 737
XM_005266023.1 XP_005266080.1 2 455
XM_011534745.1 XP_011533047.1 2 559
XM_011534746.2 XP_011533048.1 2 456
XM_011534747.1 XP_011533049.1 2 614
GenBank mRNA AF123757.1 - 2 543
AF123758.1 - 2 543
AF123759.1 - 2 543
AF123760.1 - 2 543
AF123761.1 - 2 543
AK312486.1 - 2 557
BC007725.2 - 2 468 4303103
BT007049.1 - 1 309

View Full Product Details