Gene Transcripts
- Gene Symbol
- ABLIM3
- Entrez Gene ID
- 22885
- Gene Name
- actin binding LIM protein family member 3
- Gene Aliases
- HMFN1661
- Chromosome Location
- Chr. 5: 149141483 - 149260439 on Build GRCh38
- UniGene ID
- Hs.49688
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301015.1 |
NP_001287944.1
|
8, 9 |
1437 |
|
NM_001301018.1 |
NP_001287947.1
|
8, 9 |
911 |
|
NM_001301027.1 |
NP_001287956.1
|
9, 10 |
1516 |
|
NM_001301028.1 |
NP_001287957.1
|
8, 9 |
1437 |
|
NM_014945.3 |
NP_055760.1
|
9, 10 |
1083 |
|
XM_005268392.1 |
XP_005268449.1
|
9, 10 |
1064 |
|
XM_005268393.1 |
XP_005268450.1
|
9, 10 |
1064 |
|
XM_005268394.1 |
XP_005268451.1
|
9, 10 |
1064 |
|
XM_005268395.1 |
XP_005268452.1
|
9, 10 |
1064 |
|
XM_005268396.1 |
XP_005268453.1
|
9, 10 |
1064 |
|
XM_011537609.2 |
XP_011535911.1
|
9, 10 |
1064 |
|
XM_017009238.1 |
XP_016864727.1
|
8, 9 |
1158 |
|
GenBank mRNA |
AB020650.1 |
- |
9, 10 |
996 |
|
AK225769.1 |
- |
8, 9 |
1305 |
|
AK289736.1 |
- |
8, 9 |
850 |
|
AL833021.1 |
- |
5, 6 |
476 |
|
BC001665.2 |
- |
8, 9 |
1099 |
2988043 |
DQ413174.1 |
- |
8, 9 |
873 |
|
View Full Product Details