Gene Transcripts

Gene Symbol
KAZN
Entrez Gene ID
23254
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
KAZ
Chromosome Location
Chr. 1: 13893387 - 15118048 on Build GRCh38
UniGene ID
Hs.368823
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017999.2 NP_001017999.1 5 907
NM_001018000.3 NP_001018000.1 5 1096
NM_001018001.2 NP_001018001.1 5 897
NM_015209.2 NP_056024.1 5 1159
NM_201628.2 NP_963922.2 5 1159
XM_005245795.4 XP_005245852.1 7 1414
XM_011541074.2 XP_011539376.1 7 1421
XM_011541075.2 XP_011539377.1 5 1077
XM_011541076.2 XP_011539378.1 5 1222
XM_011541077.2 XP_011539379.1 5 2201
XM_011541080.2 XP_011539382.1 7 1414
XM_017000768.1 XP_016856257.1 7 1414
XM_017000769.1 XP_016856258.1 7 1414
XM_017000770.1 XP_016856259.1 7 1414
XM_017000771.1 XP_016856260.1 6 1082
XM_017000772.1 XP_016856261.1 5 761
GenBank mRNA AB015329.1 - 4 554
AY505119.1 - 6 1087
AY505120.1 - 5 1096
AY505121.1 - 5 897
AY505122.1 - 5 907
BC035501.2 - 5 1095 4556194

View Full Product Details