Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152251.3 | NP_689464.1 | 2 | 274 | |
GenBank mRNA | AY122466.1 | - | 2 | 301 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040704.1 | NP_001035794.1 | 2 | 274 |