Gene Transcripts

Gene Symbol
BRI3
Entrez Gene ID
25798
Gene Name
brain protein I3
Gene Aliases
I3
Chromosome Location
Chr. 7: 98281667 - 98322398 on Build GRCh38
UniGene ID
Hs.567438
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015379.4 NP_056194.1 3 755
GenBank mRNA AF041430.2 - 3 725
AF106966.1 - 3 706
BC062370.1 - 3 719 6598528

View Full Product Details