Gene Transcripts

Gene Symbol
C16orf54
Entrez Gene ID
283897
Gene Name
chromosome 16 open reading frame 54
Gene Aliases
Chromosome Location
Chr. 16: 29742463 - 29746019 on Build GRCh38
UniGene ID
Hs.331095
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175900.3 NP_787096.2 2 1652
GenBank mRNA AK093000.1 - 2 1639

View Full Product Details