Gene Transcripts

Gene Symbol
SIGLEC15
Entrez Gene ID
284266
Gene Name
sialic acid binding Ig like lectin 15
Gene Aliases
CD33L3, HsT1361, SIGLEC-15
Chromosome Location
Chr. 18: 45825580 - 45842556 on Build GRCh38
UniGene ID
Hs.287692
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213602.2 NP_998767.1 6 1462
GenBank mRNA AK025833.1 - 1 109
AK092746.1 - 3 1013
AK095432.1 - 7 1166
AK172835.1 - 6 1462
AY971516.1 - 6 1451

View Full Product Details