Gene Transcripts
- Gene Symbol
- ARHGEF15
- Entrez Gene ID
- 22899
- Gene Name
- Rho guanine nucleotide exchange factor 15
- Gene Aliases
- ARGEF15, E5, Ephexin5, Vsm-RhoGEF
- Chromosome Location
- Chr. 17: 8310238 - 8322516 on Build GRCh38
- UniGene ID
- Hs.443109
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025014.1 |
NP_079290.1
|
5, 6 |
1144 |
|
NM_173728.3 |
NP_776089.2
|
5, 6 |
1208 |
|
XM_011523734.1 |
XP_011522036.1
|
5, 6 |
1208 |
|
XM_011523735.1 |
XP_011522037.1
|
5, 6 |
1144 |
|
XM_011523736.1 |
XP_011522038.1
|
5, 6 |
1208 |
|
XM_011523737.1 |
XP_011522039.1
|
2, 3 |
200 |
|
GenBank mRNA |
AB020722.1 |
- |
5, 6 |
1124 |
|
AK291626.1 |
- |
5, 6 |
1168 |
|
AK300327.1 |
- |
5, 6 |
629 |
|
BC036749.1 |
- |
5, 6 |
1163 |
5204409 |
KU178557.1 |
- |
4, 5 |
1053 |
|
KU178558.1 |
- |
4, 5 |
1053 |
|
View Full Product Details