Gene Transcripts

Gene Symbol
MRGPRG
Entrez Gene ID
386746
Gene Name
MAS related GPR family member G
Gene Aliases
GPR169, MRGG
Chromosome Location
Chr. 11: 3217944 - 3218813 on Build GRCh38
UniGene ID
Hs.730306
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164377.1 NP_001157849.1 1 46
GenBank mRNA AY255583.1 - 1 46

View Full Product Details