Gene Transcripts

Gene Symbol
GRIFIN
Entrez Gene ID
402635
Gene Name
galectin-related inter-fiber protein
Gene Aliases
Chromosome Location
Chr. 7: 2473576 - 2476855 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291784.1 NP_001278713.1 3 331
XM_011515405.2 XP_011513707.1 4 511

View Full Product Details