Gene Transcripts
- Gene Symbol
- RPL26L1
- Entrez Gene ID
- 51121
- Gene Name
- ribosomal protein L26 like 1
- Gene Aliases
- RPL26P1
- Chromosome Location
- Chr. 5: 172953677 - 172969771 on Build GRCh38
- UniGene ID
- Hs.546390
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317980.1 |
NP_001304909.1
|
2, 3 |
283 |
|
NM_001317981.1 |
NP_001304910.1
|
2, 3 |
257 |
|
NM_001317982.1 |
NP_001304911.1
|
2, 3 |
239 |
|
NM_016093.3 |
NP_057177.1
|
2, 3 |
229 |
|
XM_005265921.3 |
XP_005265978.1
|
2, 3 |
280 |
|
XM_011534565.2 |
XP_011532867.1
|
2, 3 |
844 |
|
XM_017009519.1 |
XP_016865008.1
|
2, 3 |
271 |
|
XM_017009520.1 |
XP_016865009.1
|
2, 3 |
202 |
|
GenBank mRNA |
AF083248.1 |
- |
2, 3 |
203 |
|
AK128872.1 |
- |
1 |
197 |
|
BC017360.2 |
- |
2, 3 |
217 |
4551093 |
BC070192.1 |
- |
2, 3 |
202 |
6727802 |
BI838595.1 |
- |
|
257 |
|
BQ278240.1 |
- |
2, 3 |
230 |
|
BU193755.1 |
- |
2, 3 |
263 |
|
BU569345.1 |
- |
2, 3 |
242 |
|
View Full Product Details