Gene Transcripts

Gene Symbol
LUC7L3
Entrez Gene ID
51747
Gene Name
LUC7 like 3 pre-mRNA splicing factor
Gene Aliases
CRA, CREAP-1, CROP, LUC7A, OA48-18, hLuc7A
Chromosome Location
Chr. 17: 50719565 - 50754269 on Build GRCh38
UniGene ID
Hs.130293
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006107.3 NP_006098.2 5 566
NM_016424.4 NP_057508.2 5 566
XM_005257448.1 XP_005257505.1 5 534
XM_005257449.2 XP_005257506.1 5 534
XM_005257452.1 XP_005257509.1 5 534
XM_006721943.2 XP_006722006.1 5 313
GenBank mRNA AK001925.1 - 5 500
AK023672.1 - 5 528
AK225980.1 - 5 534
AK290540.1 - 5 533
AK295983.1 - 4 435
BC002925.2 - 5 507
BC047043.1 - 5 515
BC056409.1 - 7 769
DQ013876.1 - 5 502

View Full Product Details