Gene Transcripts
- Gene Symbol
- CHIC1
- Entrez Gene ID
- 53344
- Gene Name
- cysteine rich hydrophobic domain 1
- Gene Aliases
- BRX
- Chromosome Location
- Chr. X: 73563045 - 73687109 on Build GRCh38
- UniGene ID
- Hs.496323
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039840.2 |
NP_001034929.2
|
3, 4 |
654 |
|
NM_001300884.1 |
NP_001287813.1
|
3, 4 |
654 |
|
XM_017029583.1 |
XP_016885072.1
|
4, 5 |
335 |
|
GenBank mRNA |
AK297768.1 |
- |
3, 4 |
511 |
|
AL832095.1 |
- |
3, 4 |
294 |
|
BC127727.1 |
- |
3, 4 |
565 |
40127524 |
BC127728.1 |
- |
3, 4 |
565 |
40127525 |
BC136691.1 |
- |
3, 4 |
559 |
9020681 |
BC136692.1 |
- |
3, 4 |
559 |
9020682 |
CR936642.1 |
- |
3, 4 |
605 |
|
View Full Product Details