Gene Transcripts

Gene Symbol
SDHAF1
Entrez Gene ID
644096
Gene Name
succinate dehydrogenase complex assembly factor 1
Gene Aliases
LYRM8
Chromosome Location
Chr. 19: 35995188 - 35996318 on Build GRCh38
UniGene ID
Hs.356460
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042631.2 NP_001036096.1 1 1003
GenBank mRNA BC007205.1 - 1 992

View Full Product Details