Gene Transcripts

Gene Symbol
DNLZ
Entrez Gene ID
728489
Gene Name
DNL-type zinc finger
Gene Aliases
C9orf151, HEP, HEP1, TIMM15, ZIM17, bA413M3.2
Chromosome Location
Chr. 9: 136361900 - 136363811 on Build GRCh38
UniGene ID
Hs.528581
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080849.2 NP_001074318.1 2 366
NR_073565.1 2 370
GenBank mRNA BC146915.1 - 2 366
BC146924.1 - 2 366 9056852

View Full Product Details