Gene Transcripts

Gene Symbol
PRSS46
Entrez Gene ID
100287362
Gene Name
protease, serine 46
Gene Aliases
TESSP6
Chromosome Location
Chr. 3: 46719583 - 46736431 on Build GRCh38
UniGene ID
Hs.673244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205271.1 NP_001192200.1 3 492
GenBank mRNA DB058077.1 - 3 405
DB099248.1 - 3 488
DY654973.1 - 1 174

View Full Product Details