Gene Transcripts

Gene Symbol
SORCS3
Entrez Gene ID
22986
Gene Name
sortilin related VPS10 domain containing receptor 3
Gene Aliases
SORCS
Chromosome Location
Chr. 10: 104641101 - 105265242 on Build GRCh38
UniGene ID
Hs.671950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014978.2 NP_055793.1 8 1474
XM_011539542.2 XP_011537844.1 3 225
GenBank mRNA AB028982.1 - 8 1474
BC117417.1 - 8 1290 40125968

View Full Product Details