Gene Transcripts
- Gene Symbol
- UFD1L
- Entrez Gene ID
- 7353
- Gene Name
- ubiquitin fusion degradation 1 like (yeast)
- Gene Aliases
- UFD1
- Chromosome Location
- Chr. 22: 19449941 - 19479215 on Build GRCh38
- UniGene ID
- Hs.474213
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001035247.2 |
NP_001030324.2
|
7, 8 |
695 |
|
NM_005659.6 |
NP_005650.2
|
7, 8 |
695 |
|
XM_011530361.2 |
XP_011528663.1
|
7, 8 |
821 |
|
GenBank mRNA |
AF141201.1 |
- |
6, 7 |
565 |
|
AJ239058.1 |
- |
6, 7 |
565 |
|
AK128120.1 |
- |
2, 3 |
2586 |
|
AK225877.1 |
- |
7, 8 |
652 |
|
AK304734.1 |
- |
7, 8 |
665 |
|
AK315998.1 |
- |
7, 8 |
646 |
|
AY101594.1 |
- |
7, 8 |
622 |
|
BC001049.2 |
- |
7, 8 |
642 |
3507963 |
BC005087.1 |
- |
7, 8 |
643 |
3830774 |
BM761958.1 |
- |
6, 7 |
634 |
|
BX648483.1 |
- |
7, 8 |
627 |
|
CR456607.1 |
- |
7, 8 |
579 |
|
U64444.1 |
- |
7, 8 |
751 |
|
View Full Product Details