Gene Transcripts
- Gene Symbol
- LLPH
- Entrez Gene ID
- 84298
- Gene Name
- LLP homolog, long-term synaptic facilitation
- Gene Aliases
- C12orf31, hLLP
- Chromosome Location
- Chr. 12: 66123069 - 66130753 on Build GRCh38
- UniGene ID
- Hs.504820
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091568.1 |
- |
3 |
1166 |
|
View Full Product Details