Gene Transcripts

Gene Symbol
UNC13A
Entrez Gene ID
23025
Gene Name
unc-13 homolog A
Gene Aliases
Munc13-1
Chromosome Location
Chr. 19: 17601328 - 17688344 on Build GRCh38
UniGene ID
Hs.164502
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080421.2 NP_001073890.2 24 2932
XM_011527810.2 XP_011526112.1 23 3047
XM_011527811.2 XP_011526113.1 24 3077
XM_017026502.1 XP_016881991.1 24 3077
GenBank mRNA AB028955.2 - 24 2930

View Full Product Details