Gene Transcripts

Gene Symbol
SAMD4A
Entrez Gene ID
23034
Gene Name
sterile alpha motif domain containing 4A
Gene Aliases
SAMD4, SMAUG, SMAUG1, SMG, SMGA
Chromosome Location
Chr. 14: 54567612 - 54793315 on Build GRCh38
UniGene ID
Hs.733324
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015589.5 NP_056404.4 3 1048
GenBank mRNA AK024652.1 - 3 741

View Full Product Details