Gene Transcripts

Gene Symbol
SAMD4A
Entrez Gene ID
23034
Gene Name
sterile alpha motif domain containing 4A
Gene Aliases
SAMD4, SMAUG, SMAUG1, SMG, SMGA
Chromosome Location
Chr. 14: 54567612 - 54793315 on Build GRCh38
UniGene ID
Hs.733324
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161576.2 NP_001155048.2 7 1745
NM_001161577.1 NP_001155049.1 4 638
NM_015589.5 NP_056404.4 8 2009
GenBank mRNA AB028976.1 - 6 1076
AK024652.1 - 8 1702
AK025923.1 - 6 900
BC021060.1 - 4 555
BC021726.2 - 4 593
BC121173.1 - 7 1442 40122465
BC121174.1 - 7 1442 40122467
BI825501.1 - 4 638

View Full Product Details