Gene Transcripts

Gene Symbol
RAB1A
Entrez Gene ID
5861
Gene Name
RAB1A, member RAS oncogene family
Gene Aliases
RAB1, YPT1
Chromosome Location
Chr. 2: 65086854 - 65130301 on Build GRCh38
UniGene ID
Hs.310645
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004161.4 NP_004152.1 3 554
NM_015543.1 NP_056358.1 3 554
XM_005264468.1 XP_005264525.1 3 359
GenBank mRNA AF498929.1 - 3 168
AK303678.1 - 3 358
AK315495.1 - 3 363
AL050268.1 - 3 358
BC000905.2 - 3 346 2900705
BG704216.1 - 3 668
BT019528.1 - 3 168
BT019529.1 - 3 168
BX571747.1 - 3 351
CR533479.1 - 3 168
CR536488.1 - 3 168
M28209.1 - 3 218
X13898.1 - 3 168

View Full Product Details