Gene Transcripts

Gene Symbol
C9orf57
Entrez Gene ID
138240
Gene Name
chromosome 9 open reading frame 57
Gene Aliases
Chromosome Location
Chr. 9: 72051381 - 72060605 on Build GRCh38
UniGene ID
Hs.371235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128618.1 NP_001122090.1 3, 4 326
XM_011518225.2 XP_011516527.1 2, 3 170
GenBank mRNA BC036255.1 - 3, 4 326
BC044620.1 - 3, 4 304
BC053956.1 - 3, 4 304
BC130404.1 - 2, 3 225 40146435
BC144273.1 - 2, 3 224 9052793

View Full Product Details