Gene Transcripts

Gene Symbol
WAPL
Entrez Gene ID
23063
Gene Name
WAPL cohesin release factor
Gene Aliases
FOE, KIAA0261, WAPAL
Chromosome Location
Chr. 10: 86435256 - 86521816 on Build GRCh38
UniGene ID
Hs.203099
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318328.1 NP_001305257.1 10 2835
NM_015045.3 NP_055860.1 10 2853
XM_011539548.1 XP_011537850.1 10 2903
XM_017015979.1 XP_016871468.1 9 2750
XM_017015980.1 XP_016871469.1 9 2693
XM_017015981.1 XP_016871470.1 7 2440
GenBank mRNA AB065003.1 - 10 2856
AF479418.1 - 11 2531
AK027005.1 - 3 576
BC017393.1 - 3 289
BC140858.1 - 10 2559 9021726
BC150269.1 - 11 2704 9007232
D87450.1 - 11 2704

View Full Product Details