Gene Transcripts

Gene Symbol
JAKMIP3
Entrez Gene ID
282973
Gene Name
Janus kinase and microtubule interacting protein 3
Gene Aliases
C10orf14, C10orf39, Jamip3, NECC2, bA140A10.5
Chromosome Location
Chr. 10: 132036068 - 132184852 on Build GRCh38
UniGene ID
Hs.106254
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105521.3 NP_001098991.1 3 551
NM_001323086.1 NP_001310015.1 3 551
NM_001323087.1 NP_001310016.1 3 551
NM_001323088.1 NP_001310017.1 3 551
NM_001323089.1 NP_001310018.1 3 551
NM_001323090.1 NP_001310019.1 3 551
XM_005252674.4 XP_005252731.1 3 2545
XM_005252676.4 XP_005252733.1 3 2545
XM_011539676.2 XP_011537978.1 3 797
XM_011539682.2 XP_011537984.1 3 2545
XM_017016095.1 XP_016871584.1 3 2545
XM_017016096.1 XP_016871585.1 3 796
XM_017016097.1 XP_016871586.1 3 2545
XM_017016098.1 XP_016871587.1 3 2545
XM_017016099.1 XP_016871588.1 3 2545

View Full Product Details