Gene Transcripts

Gene Symbol
PATE3
Entrez Gene ID
100169851
Gene Name
prostate and testis expressed 3
Gene Aliases
HEL-127, PATE-DJ
Chromosome Location
Chr. 11: 125785996 - 125791600 on Build GRCh38
UniGene ID
Hs.131278
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129883.3 NP_001123355.3 3 289
XM_017017054.1 XP_016872543.1 2 3589
GenBank mRNA EF426753.1 - 3 289
EU703625.1 - 3 248

View Full Product Details