Gene Transcripts
- Gene Symbol
- NBPF1
- Entrez Gene ID
- 55672
- Gene Name
- neuroblastoma breakpoint family member 1
- Gene Aliases
- AB13, AB14, AB23, AD2, NBG, NBPF
- Chromosome Location
- Chr. 1: 16562427 - 16613605 on Build GRCh38
- UniGene ID
- Hs.467587
- Species
- Homo sapiens
- Gene Symbol
- LOC102724250
- Entrez Gene ID
- 102724250
- Gene Name
- neuroblastoma breakpoint family member 1
- Gene Aliases
- Chromosome Location
- Chr. 1|NT_187366: 4611 - 41174 on Build GRCh38
- UniGene ID
- Hs.650616
- Species
- Homo sapiens
- Gene Symbol
- NBPF10
- Entrez Gene ID
- 100132406
- Gene Name
- neuroblastoma breakpoint family member 10
- Gene Aliases
- AB6, AG1, NBPF9
- Chromosome Location
- Chr. 1: 146066318 - 146144804 on Build GRCh38
- UniGene ID
- Hs.714127
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF379616.1 |
- |
9 |
1437 |
|
AF419616.1 |
- |
23 |
2822 |
|
BX538005.1 |
- |
28 |
4134 |
|
- Gene Symbol
- NBPF14
- Entrez Gene ID
- 25832
- Gene Name
- neuroblastoma breakpoint family member 14
- Gene Aliases
- DJ328E19.C1.1, NBPF
- Chromosome Location
- Chr. 1: 148531385 - 148607636 on Build GRCh38
- UniGene ID
- Hs.534675
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF379626.1 |
- |
12 |
1533 |
|
AK095459.1 |
- |
12 |
2319 |
|
- Gene Symbol
- NBPF15
- Entrez Gene ID
- 284565
- Gene Name
- neuroblastoma breakpoint family member 15
- Gene Aliases
- AB14, AG3, NBPF16
- Chromosome Location
- Chr. 1: 144421386 - 144459464 on Build GRCh38
- UniGene ID
- Hs.523572
- Species
- Homo sapiens
- Gene Symbol
- NBPF19
- Entrez Gene ID
- 101060226
- Gene Name
- neuroblastoma breakpoint family member 19
- Gene Aliases
- Chromosome Location
- Chr. 1: 149472955 - 149556256 on Build GRCh38
- UniGene ID
- Hs.445080
- Species
- Homo sapiens
- Gene Symbol
- NBPF20
- Entrez Gene ID
- 100288142
- Gene Name
- neuroblastoma breakpoint family member 20
- Gene Aliases
- Chromosome Location
- Chr. 1: 145289900 - 145405778 on Build GRCh38
- UniGene ID
- Hs.445080
- Species
- Homo sapiens
- Gene Symbol
- NBPF9
- Entrez Gene ID
- 400818
- Gene Name
- neuroblastoma breakpoint family member 9
- Gene Aliases
- AE01
- Chromosome Location
- Chr. 1: 149054033 - 149090861 on Build GRCh38
- UniGene ID
- Hs.445080
- Species
- Homo sapiens
View Full Product Details