Gene Transcripts

Gene Symbol
SPEG
Entrez Gene ID
10290
Gene Name
SPEG complex locus
Gene Aliases
APEG-1, APEG1, BPEG, CNM5, SPEGalpha, SPEGbeta
Chromosome Location
Chr. 2: 219434215 - 219493632 on Build GRCh38
UniGene ID
Hs.21639
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005876.4 NP_005867.3 30 6973
XM_005246237.2 XP_005246294.1 32 6701
XM_005246239.2 XP_005246296.1 32 6516
XM_005246240.2 XP_005246297.1 25 4718
XM_005246241.1 XP_005246298.1 25 4612
XM_005246242.4 XP_005246299.1 25 4737
XM_006712189.3 XP_006712252.1 31 6706
XM_006712193.3 XP_006712256.1 25 7837
XM_011510479.2 XP_011508781.1 31 7139
XM_011510483.2 XP_011508785.2 31 6736
XM_017003157.1 XP_016858646.1 31 6736
XM_017003158.1 XP_016858647.1 27 6312
XM_017003159.1 XP_016858648.1 28 6495
XM_017003160.1 XP_016858649.1 15 2908
GenBank mRNA AB037718.1 - 21 4030
AY603755.1 - 29 6586

View Full Product Details