Gene Transcripts

Gene Symbol
TSPAN14
Entrez Gene ID
81619
Gene Name
tetraspanin 14
Gene Aliases
DC-TM4F2, TM4SF14
Chromosome Location
Chr. 10: 80454271 - 80522635 on Build GRCh38
UniGene ID
Hs.718943
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128309.1 NP_001121781.1 3, 4 313
NM_030927.2 NP_112189.2 6, 7 682
XM_006717992.3 XP_006718055.1 6, 7 838
XM_011540219.2 XP_011538521.1 8, 9 872
XM_011540220.2 XP_011538522.1 8, 9 794
XM_011540221.2 XP_011538523.1 7, 8 656
XM_011540222.2 XP_011538524.1 7, 8 834
XM_017016707.1 XP_016872196.1 8, 9 830
XM_017016708.1 XP_016872197.1 7, 8 791
XM_017016709.1 XP_016872198.1 7, 8 839
XM_017016710.1 XP_016872199.1 7, 8 790
XM_017016711.1 XP_016872200.1 7, 8 655
XM_017016712.1 XP_016872201.1 7, 8 858
XM_017016713.1 XP_016872202.1 6, 7 678
GenBank mRNA AF311903.1 - 6, 7 647
AK295321.1 - 3, 4 284
AL136638.1 - 6, 7 653
BC002920.2 - 5, 6 577 3954042
BC093016.1 - 6, 7 682 30528940
BI918996.1 - 3, 4 336
DC300676.1 - 3, 4 285

View Full Product Details