Gene Transcripts

Gene Symbol
ZBTB43
Entrez Gene ID
23099
Gene Name
zinc finger and BTB domain containing 43
Gene Aliases
ZBTB22B, ZNF-X, ZNF297B
Chromosome Location
Chr. 9: 126804072 - 126838210 on Build GRCh38
UniGene ID
Hs.355581
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135776.1 NP_001129248.1 2 361
NM_014007.3 NP_054726.1 3 484
XM_005251835.3 XP_005251892.1 3 805
XM_011518409.1 XP_011516711.1 4 497
XM_011518411.2 XP_011516713.1 2 611
GenBank mRNA AB007874.1 - 1 1343
AF049907.1 - 3 464
AK026123.1 - 3 430
BC008828.2 - 3 408 4123095
BT007194.1 - 1 211
DA500531.1 - 2 357
DA864775.1 - 3 486

View Full Product Details