Gene Transcripts

Gene Symbol
MICAL3
Entrez Gene ID
57553
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 3
Gene Aliases
MICAL-3
Chromosome Location
Chr. 22: 17787649 - 18024559 on Build GRCh38
UniGene ID
Hs.528024
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122731.2 NP_001116203.1 7 1062
NM_001136004.3 NP_001129476.1 7 1062
NM_015241.2 NP_056056.2 8 1341
XM_005261262.3 XP_005261319.1 8 1171
XM_011546137.2 XP_011544439.1 8 1171
XM_011546139.2 XP_011544441.1 8 1171
XM_011546142.2 XP_011544444.1 8 1171
XM_011546146.2 XP_011544448.1 8 1171
XM_017028865.1 XP_016884354.1 8 1689
XM_017028866.1 XP_016884355.1 8 1495
XM_017028867.1 XP_016884356.1 8 2612
XM_017028868.1 XP_016884357.1 8 1341
XM_017028869.1 XP_016884358.1 8 1086
XM_017028870.1 XP_016884359.1 8 1087
XM_017028871.1 XP_016884360.1 8 1171
XM_017028872.1 XP_016884361.1 8 1171
XM_017028873.1 XP_016884362.1 8 1171
XM_017028874.1 XP_016884363.1 8 1171
XM_017028875.1 XP_016884364.1 8 1171
XM_017028876.1 XP_016884365.1 8 1171
XM_017028877.1 XP_016884366.1 8 1171
XM_017028878.1 XP_016884367.1 8 1171
XM_017028879.1 XP_016884368.1 8 1171
XM_017028880.1 XP_016884369.1 8 1494
XM_017028881.1 XP_016884370.1 8 1341
XM_017028882.1 XP_016884371.1 8 1171
XM_017028883.1 XP_016884372.1 7 834
XM_017028884.1 XP_016884373.1 9 1620
XM_017028887.1 XP_016884376.1 8 1171
GenBank mRNA AB037785.1 - 5 493
AK127487.1 - 7 1070
BC157876.1 - 7 1050 8862700
BC171887.1 - 7 1050 9054541
BX647382.1 - 7 1011
CR456364.1 - 7 1003

View Full Product Details