Gene Transcripts

Gene Symbol
Speer4d
Entrez Gene ID
360220
Gene Name
spermatogenesis associated glutamate (E)-rich protein 4d
Gene Aliases
4933431D05Rik, SPEER-4D
Chromosome Location
Chr. 5: 15619099 - 15623864 on Build GRCm38
UniGene ID
Mm.425235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025759.3 NP_080035.2 5 903
GenBank mRNA AK017002.1 - 5 904

View Full Product Details