Gene Transcripts

Gene Symbol
Cnfn
Entrez Gene ID
72383
Gene Name
cornifelin
Gene Aliases
2210418J09Rik
Chromosome Location
Chr. 7: 25367616 - 25369724 on Build GRCm38
UniGene ID
Mm.34382
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081375.1 NP_001074844.1 2 119
NM_028219.1 NP_082495.1 3 312
GenBank mRNA AK008979.1 - 2 120
BC059093.1 - 3 291 30293557

View Full Product Details