Gene Transcripts

Gene Symbol
Pqlc1
Entrez Gene ID
66943
Gene Name
PQ loop repeat containing 1
Gene Aliases
2310009N05Rik, 4933425L21Rik, 5730564E11Rik, C78974
Chromosome Location
Chr. 18: 80255241 - 80292724 on Build GRCm38
UniGene ID
Mm.29247
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164420.1 NP_001157892.1 3 568
NM_001164421.1 NP_001157893.1 3 500
NM_001164422.1 NP_001157894.1 3 495
NM_001164423.1 NP_001157895.1 3 500
NM_001164424.1 NP_001157896.1 3 495
NM_025861.3 NP_080137.2 3 500
GenBank mRNA AK009256.1 - 3 490
AK017854.1 - 3 508
AK018759.1 - 2 878
AK019858.1 - 3 442
AK047446.1 - 3 433
AK153023.1 - 3 520
AK153176.1 - 3 521
BC008231.1 - 3 437 2647794
BC029179.1 - 3 442 3487991
BC043686.1 - 678 5135928
CJ135868.1 - 3 520

View Full Product Details