Gene Transcripts

Gene Symbol
ZCCHC14
Entrez Gene ID
23174
Gene Name
zinc finger CCHC-type containing 14
Gene Aliases
BDG-29, BDG29
Chromosome Location
Chr. 16: 87406246 - 87492948 on Build GRCh38
UniGene ID
Hs.156231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015144.2 NP_055959.1 5 528
XM_005255858.3 XP_005255915.2 5 719
XM_011522964.2 XP_011521266.1 5 1622
XM_017023082.1 XP_016878571.1 5 1478
GenBank mRNA AB011151.2 - 5 386
AB030243.1 - 5 536
AK289682.1 - 5 719
AL117532.1 - 2 151
BC101478.1 - 5 535 8068984

View Full Product Details