Gene Transcripts

Gene Symbol
Wdr70
Entrez Gene ID
545085
Gene Name
WD repeat domain 70
Gene Aliases
4833422F06Rik
Chromosome Location
Chr. 15: 7873055 - 8099209 on Build GRCm38
UniGene ID
Mm.321312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081402.1 NP_001074871.1 7 684
GenBank mRNA AK159714.1 - 7 666
AK160933.1 - 7 619

View Full Product Details