Gene Transcripts

Gene Symbol
Cfhr2
Entrez Gene ID
545366
Gene Name
complement factor H-related 2
Gene Aliases
Cfhrb_4/2
Chromosome Location
Chr. 1: 139810292 - 139858699 on Build GRCm38
UniGene ID
Mm.439660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025575.2 NP_001020746.1 6 950
GenBank mRNA AJ565908.1 - 6 899
AK149511.1 - 6 941
BC026782.1 - 1465
M29010.1 - 1623

View Full Product Details