Gene Transcripts

Gene Symbol
Gal3st2
Entrez Gene ID
381334
Gene Name
galactose-3-O-sulfotransferase 2
Gene Aliases
GP3ST, Gm988
Chromosome Location
Chr. 1: 93861344 - 93876494 on Build GRCm38
UniGene ID
Mm.482878
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199366.4 NP_955398.3 4 1849
GenBank mRNA AK144526.1 - 4 1850
AK165692.1 - 4 1853

View Full Product Details