Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207131.1 | NP_997014.1 | 2 | 904 | |
GenBank mRNA | AK143561.1 | - | 2 | 905 | |
AY570294.1 | - | 2 | 904 | ||
BC108953.1 | - | 2 | 841 | 40045189 | |
BC108954.1 | - | 2 | 841 | 40045190 |