Gene Transcripts

Gene Symbol
R3hdml
Entrez Gene ID
100043899
Gene Name
R3H domain containing-like
Gene Aliases
OTTMUSG00000001070
Chromosome Location
Chr. 2: 163491214 - 163504364 on Build GRCm38
UniGene ID
Mm.442535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099331.2 NP_001092801.1 3 678

View Full Product Details