Gene Transcripts

Gene Symbol
Pbrm1
Entrez Gene ID
66923
Gene Name
polybromo 1
Gene Aliases
2610016F04Rik, AI507524, BAF180, Pb1
Chromosome Location
Chr. 14: 31019138 - 31121592 on Build GRCm38
UniGene ID
Mm.27913
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081251.1 NP_001074720.1 19, 20 2926
GenBank mRNA AK030252.1 - 9, 10 1717
AK030781.1 - 18, 19 2829
AK165182.1 - 18, 19 2884
AK166588.1 - 18, 19 2880
BC024438.1 - 10, 11 1782
BC029037.1 - 17, 18 2698
BC037018.1 - 5, 6 1169

View Full Product Details