Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020584.2 | NP_065609.2 | 3 | 1178 | |
GenBank mRNA | AK041613.1 | - | 3 | 1124 | |
AK051818.1 | - | 3 | 1098 | ||
AK081557.1 | - | 3 | 1134 | ||
AK084563.1 | - | 3 | 1126 | ||
AK087729.1 | - | 3 | 1136 | ||
AK148508.1 | - | 3 | 1138 | ||
AK148537.1 | - | 3 | 1140 | ||
AK161452.1 | - | 3 | 1157 | ||
AK165074.1 | - | 3 | 1136 | ||
BC012270.1 | - | 3 | 1094 | 3483521 | |
BC017641.1 | - | 3 | 1154 | 4483320 |