Gene Transcripts
- Gene Symbol
- Srp54b
- Entrez Gene ID
- 665155
- Gene Name
- signal recognition particle 54B
- Gene Aliases
- Chromosome Location
- Chr. 12: 55155104 - 55189213 on Build GRCm38
- UniGene ID
- Mm.12848
- Species
- Mus musculus
- Gene Symbol
- Srp54a
- Entrez Gene ID
- 24067
- Gene Name
- signal recognition particle 54A
- Gene Aliases
- 54kDa, Srp54
- Chromosome Location
- Chr. 12: 55080496 - 55115367 on Build GRCm38
- UniGene ID
- Mm.471968
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011899.4 |
NP_036029.2
|
16 |
2687 |
|
View Full Product Details