Gene Transcripts

Gene Symbol
Cypt1
Entrez Gene ID
66742
Gene Name
cysteine-rich perinuclear theca 1
Gene Aliases
4931420D14Rik, Ckt1r3, Cypt5, Cypt6, TISP55
Chromosome Location
Chr. X: 16522855 - 16523691 on Build GRCm38
UniGene ID
Mm.45302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025738.2 NP_080014.2 2 642
GenBank mRNA AK006892.1 - 2 630
AK016467.1 - 2 644
BC048455.1 - 2 626
BC061079.1 - 2 626 6773161

View Full Product Details