Gene Transcripts

Gene Symbol
Bhlhe22
Entrez Gene ID
59058
Gene Name
basic helix-loop-helix family, member e22
Gene Aliases
Beta3, Bhlhb5
Chromosome Location
Chr. 3: 18054325 - 18057514 on Build GRCm38
UniGene ID
Mm.149938
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021560.4 NP_067535.3 1 1670
GenBank mRNA AF218865.1 - 1 1292
AK047201.1 - 1 1404
BC053007.1 - 1 1670 5686844

View Full Product Details